missing translation for 'onlineSavingsMsg'
Learn More

Carboxymethoxylamine HemiHydroChloride, 98%

Numéro de catalogue. AC108430100
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC108430100 10 g Glass bottle
AC108430025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC108430100 Fournisseur Thermo Scientific Chemicals Code fournisseur 108430100
Il en reste null

CAS: 2921-14-4 | C4H11ClN2O6 | 218.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 2921-14-4
Molecular Formula C4H11ClN2O6
Molecular Weight (g/mol) 218.59
MDL Number MFCD00012955
InChI Key AXIKLJPWEGNZIG-UHFFFAOYSA-N
Synonym o-carboxymethyl hydroxylamine hemihydrochloride, carboxymethoxylamine hemihydrochloride, aminooxyacetic acid hemihydrochloride, 2-aminooxy acetic acid hydrochloride 2:1, unii-5g5qi2gn7a, acetic acid, aminooxy-, hydrochloride 2:1, aminooxy acetic acid hemihydrochloride, 5g5qi2gn7a, carboxymethoxyamine hemihydrochloride
PubChem CID 2723609
IUPAC Name 2-aminooxyacetic acid;hydrochloride
SMILES [Cl-].NOCC(O)=O.[NH3+]OCC(O)=O

Spécifications

Melting Point 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2NOCH2CO2H)2·HCl
Quantity 10 g
Solubility Information Solubility in water: decomposes.
Formula Weight 109.3
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Carboxymethoxylamine hemihydrochloride
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 220-862-3

RUO – Research Use Only