missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Carbotrap™ 300 Thermal Desorption Tubes

Suitable for use with Air Monitoring

Supplier:  MilliporeSigma™ Supelco™ 29751U

 View more versions of this product

Catalog No. 111005344


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10/Pk.
Carbosieve™ S-III Carbon Molecular Sieve (CMS), Carbotrap™ 300, Carbotrap™ B Graphitized Carbon Black (GCB), Carbotrap™ C Graphitized Carbon Black (GCB)
Thermal Desorption Tube
Glass
PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
EPA TO-1,TO-2,TO-3,TO-17
Pre-conditioned/Sealed with Brass Endcaps
Air Monitoring
1/4 in.
Carbotrap
3.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC