Learn More
MilliporeSigma™ Supelco™ Carbotrap™ 217 Thermal Desorption Tubes
Suitable for use with Air Monitoring
Supplier:  MilliporeSigma™ Supelco™ 29750U
 View more versions of this product
Catalog No. 111005347
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
10/Pk.
Carbotrap™ 217, Carbotrap™ B Graphitized Carbon Black (GCB), Carboxen™ 1000 Carbon Molecular Sieve (CMS)
1/4 in.
Carbotrap
3.5 in.
Pre-conditioned/Sealed with Brass Endcaps
Air Monitoring
Glass
PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
EPA TO-14, EPA TO-17
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC