missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Carbotrap™ 217 Thermal Desorption Tubes

Catalog No. p-7202940
Change view
Click to view available options
Material:
Glass
Stainless Steel
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
Unconditioned
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Description
111005378 Glass Unconditioned
111005360 Glass Pre-conditioned
111005347 Glass Pre-conditioned/Sealed with Brass Endcaps
111007945 Stainless Steel Unconditioned
111005355 Stainless Steel Pre-conditioned
111005346 Stainless Steel Pre-conditioned/Sealed with Brass Endcaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 111005378 Supplier MilliporeSigma™ Supelco™ Supplier No. 30138U
Only null left

Suitable for use with Air Monitoring

Specifications

Description Unconditioned
Overall Length (English) 3.5 in.
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Diameter (English) Outer 1/4 in.
Product Line Carbotrap
For Use With (Application) Air Monitoring
Packaging Quantity 10/Pk.
Matrix Carbotrap™ 217, Carbotrap™ B Graphitized Carbon Black (GCB), Carboxen™ 1000 Carbon Molecular Sieve (CMS)
Material Glass
Methods EPA TO-14, EPA TO-17

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC