Learn More
MilliporeSigma™ Supelco™ Carbopack™ B Thermal Desorption Tubes
Use for the trapping and extraction of herbicides, simazine, and atrazine in water samples followed by high-performance liquid chromatography (HPLC) analysis
Supplier:  MilliporeSigma™ Supelco™ 29535U
 View more versions of this product
Catalog No. 111005367
Only null left
Explore available promotions
Description
Description
  • Surface Area: 100 m2/g
  • USP Code: S12
Specifications
Specifications
10/Pk.
Carbopack™ Graphitized Carbon Black (GCB)
Thermal Sorption Tube
Glass
PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Pre-conditioned
Air Monitoring, Sampling a Wide Range of VOCs
1/4 in.
Carbopack
3 1/2 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Carbopack is a trademark of Sigma-Aldrich Co. LLC