Learn More
Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min
Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation | CAS: 17185-29-4 | C55H46OP3Rh | 918.80 g/mol
$96.37 - $540.18
Chemical Identifiers
| CAS | 17185-29-4 |
|---|---|
| Molecular Formula | C55H46OP3Rh |
| Molecular Weight (g/mol) | 918.80 |
| MDL Number | MFCD00151644 |
| InChI Key | AACIZACVKFEETJ-UHFFFAOYSA-N |
| Synonym | tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i |
| PubChem CID | 132451836 |
| IUPAC Name | carbon monoxide;rhodium;triphenylphosphane |
| SMILES | [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Description
Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 17185-29-4 | |
| 918.80 | |
| AACIZACVKFEETJ-UHFFFAOYSA-N | |
| 132451836 | |
| [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
| C55H46OP3Rh | |
| MFCD00151644 | |
| tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i | |
| carbon monoxide;rhodium;triphenylphosphane |
Specifications
| 17185-29-4 | |
| Yellow | |
| Odorless | |
| RhH(CO)[P(C6H5)3]3 | |
| 0.1 g | |
| 14,9758 | |
| Soluble in hydrocarbons (e.g. benzene and toluene) with dissociation of phosphine ligands | |
| [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 918.80 | |
| 918.8 | |
| Powder |
| 138°C (decomposition) | |
| 1.33 g/mL | |
| C55H46OP3Rh | |
| MFCD00151644 | |
| Moisture sensitive | |
| tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i | |
| AACIZACVKFEETJ-UHFFFAOYSA-N | |
| carbon monoxide;rhodium;triphenylphosphane | |
| 132451836 | |
| Rh 10.0% min. | |
| Carbonylhydridotris(triphenylphosphine)rhodium(I) |
Safety and Handling
P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 241-230-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only