accessibility menu, dialog, popup

UserName

CAS RN 17185-29-4

IUPAC Name: methanidylidyneoxidanium tris(triphenylphosphane) λ¹-rhodium
Synonyms: carbon monoxide tris(triphenylphosphine) λ¹-rhodium
Molecular Weight (g/mol): 918.8
Molecular Formula: C55H46OP3Rh
InChi Key: AACIZACVKFEETJ-UHFFFAOYSA-N
SMILES: [RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results