missing translation for 'onlineSavingsMsg'
Learn More

Carbonylhydridotris(triphenylphosphine)rhodium(I), Rh 10.0% min

Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation | CAS: 17185-29-4 | C55H46OP3Rh | 918.80 g/mol

Supplier:  Thermo Scientific Chemicals 01001603

 View more versions of this product

Catalog No. AA1001603

Only null left
Explore available promotions

Description

Description

Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Carbonylhydridotris(triphenylphosphine)rhodium(I)
138°C (decomposition)
1.33 g/mL
C55H46OP3Rh
MFCD00151644
Moisture sensitive
tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i
AACIZACVKFEETJ-UHFFFAOYSA-N
carbon monoxide;rhodium;triphenylphosphane
132451836
Rh 10.0% min.
17185-29-4
Yellow
Odorless
RhH(CO)[P(C6H5)3]3
1 g
14,9758
Soluble in hydrocarbons (e.g. benzene and toluene) with dissociation of phosphine ligands
[RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
918.80
918.8
Powder

Chemical Identifiers

17185-29-4
918.80
AACIZACVKFEETJ-UHFFFAOYSA-N
132451836
[RhH].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C55H46OP3Rh
MFCD00151644
tristriphenylphosphine rhodium carbonyl hydride, hydridocarbonyltris triphenylphosphine rhodium i
carbon monoxide;rhodium;triphenylphosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c

H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 241-230-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only