missing translation for 'onlineSavingsMsg'
Learn More

Carbon Standard, (Sodium Carbonate-Sodium Bicarbonate), Certified, 1000ppm Inorganic (1mL = 1mg C), LabChem™

Supplier:  LabChem LC129001

 View more versions of this product

Catalog No. LC129001


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Carbon Standard
Poly Bottle
99.21,0.44,0.35
1g/mL
CHNaO3
Na2CO3
Colorless
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium hydrogen carbonate
500 mL
CHEBI:32139
Certified
1000ppm, Inorganic (1mL = 1mg C), (Sodium Carbonate-Sodium Bicarbonate)
144-55-8 , 497-19-8 , 144-55-8 , 7732-18-5
Passes Test
Liquid
1000 ppm Inorganic (1mL = 1mg C)
MFCD00003528
Soluble in water
[Na+].OC([O-])=O
84.01
516892
105.99

Chemical Identifiers

144-55-8
84.01
UIIMBOGNXHQVGW-UHFFFAOYSA-M
CHEBI:32139
[Na+].OC([O-])=O
CHNaO3
MFCD00003528
516892
sodium hydrogen carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions