missing translation for 'onlineSavingsMsg'
Learn More

Carbon Standard, (Sodium Carbonate-Sodium Bicarbonate), Certified, 1000ppm Inorganic (1mL = 1mg C), LabChem™

Catalog No. LC129007
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC129007 125 mL Poly Bottle
LC129001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC129007 Supplier LabChem Supplier No. LC129007

Chemical Identifiers

CAS 144-55-8 , 497-19-8 , 144-55-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 99.21, 0.44, 0.35
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 1000 ppm Inorganic (1mL = 1mg C)
Linear Formula Na2CO3
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 105.99
Grade Certified
Chemical Name or Material Carbon Standard, 1000ppm, Inorganic (1mL = 1mg C), (Sodium Carbonate-Sodium Bicarbonate)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature