missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 1000 ppm C, Ricca Chemical

Catalog No. RC18474
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
10 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RC18474 120 mL Amber Glass Bottle
184716 500 mL Amber Glass Bottle
184732 1 L Amber Glass Bottle
184725 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RC18474 Supplier Ricca Chemical Company Supplier No. 18474

Chemical Identifiers

CAS 877-24-7 , 7664-38-2 , 877-24-7 , 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 1000ppm
CAS Min % 97.0, 0.46, 0.21
CAS Max % 100.0, 0.48, 0.22
Equivalence 1 mL = 1 mg C
Physical Form Liquid
Color Colorless
pH < 2
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Organic Carbon Standard
Show More Show Less