missing translation for 'onlineSavingsMsg'
Learn More

Carbon Nanotube Single-walled (>85%) below 3nm(Average diam.), over 5micro m(Average length), TCI America™

Catalog No. C3133200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3133200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3133200MG Supplier TCI America Supplier No. C3133200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 308068-56-6
Molecular Formula CH4
Molecular Weight (g/mol) 16.043
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
PubChem CID 297
ChEBI CHEBI:16183
IUPAC Name methane
SMILES C

Specifications

Color Black
Quantity 200 mg
Physical Form Powder
Chemical Name or Material Carbon Nanotube Single-walled (>85%) below 3nm(Average diam.), over 5micro m(Average length)