missing translation for 'onlineSavingsMsg'
Learn More

Carbon Nanotube Aligned Multi-walled 10-20nm(diam.), 5-15micro m(length), TCI America™

Catalog No. C21511G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C21511G Supplier TCI America Supplier No. C21511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 308068-56-6
Molecular Formula CH4
Molecular Weight (g/mol) 16.043
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
PubChem CID 297
ChEBI CHEBI:16183
IUPAC Name methane
SMILES C

Specifications

Color Black
Quantity 1 g
Physical Form Powder
Chemical Name or Material Carbon Nanotube Aligned Multi-walled 10-20nm(diam.), 5-15micro m(length)