missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, ACS, 99.9+%

Catalog No. AA39785K3
Change view
Click to view available options
Quantity:
2 L
4 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39785K3 2 L
AA39785AY 4 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39785K3 Supplier Thermo Scientific Chemicals Supplier No. 039785K3

For Use in Electronic Vacuum Tubes, in the Manufacturing of Carbon Tetrachloride, Rayon | CAS: 75-15-0 | CS2 | 76.131 g/mol

It is used in electronic vacuum tubes, in the manufacturing of carbon tetrachloride, rayon, soil disinfectants, as a solvent, manufacture of rayon, cellophane, and carbon tetrachloride, to produce rubber and chemicals. It is used in the manufacture of xanthates. It is also used as a reaction agent for the production of rubber vulcanization. accelerators and soil sterilizers. It is an effective solvent for oils, waxes, sulfur and sulfur compounds. Carbon disulfide and carbon dioxide acts as resources for functional polymers. It is also useful for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Evaporation Residue 0.002%, Hydrogen sulfide (H2S) P.T. (limit about 1.5ppm, Sulfur dioxide (SO2) P.T. (limit about 2.5ppm), H2O 0.05%

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point °C to 112°C
Color Colorless
Appearance Colorless liquid
Density 1.263 g/cm3 at 20°C
Quantity 2 L
Flash Point -30°C (-22°F)
Ignition Point 95°C (203°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.627
Assay Percent Range >99.9%
Vapor Pressure 400 hPa (300mm Hg) at 20°C
Boiling Point 46°C
Solubility Information Soluble in alcohol,ether,benzene,oil,chloroform and carbon tetrachloride.
Formula Weight 76.14
Grade ACS
Chemical Name or Material Carbon disulfide, For analysis APS >5 micron
Show More Show Less
Hazard Category H225-H315-H319-H332-H336-H361fd-H372
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: I
EINECSNumber 200-843-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only