missing translation for 'onlineSavingsMsg'
Learn More

Carbon Dioxide ISA, Fisherbrand

Catalog No. ACCU0805500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0805500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0805500 Supplier Ricca Chemical Company Supplier No. ACCU0805500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6132-04-3 , 7647-01-0 , 7732-18-5
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
Solubility Miscible with water
Solution Type Calibration Solution
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Carbon Dioxide ISA
Show More Show Less