missing translation for 'onlineSavingsMsg'
Learn More

CO2 Activity Standard, 0.100 M Sodium Bicarbonate, Fisherbrand

Catalog No. ACCU0804500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0804500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0804500 Supplier Ricca Chemical Company Supplier No. ACCU0804500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144-55-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-component Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Carbon Dioxide Activity Standard, 0.1M Sodium Bicarbonate
Show More Show Less