missing translation for 'onlineSavingsMsg'
Learn More

Carbon black, acetylene, 50% compressed, 99.9+%

Numéro de catalogue. p-4408792
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA3972430 250 g
AA39724A1 1 kg
AA39724A5 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AA3972430 Fournisseur Thermo Scientific Chemicals Code fournisseur 03972430
Il en reste null

Form:S.A. 75m2/g, Bulk density 80-120g/L | CAS: 1333-86-4 | C | 12.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N

Spécifications

Color Black
Density Bulk density 80 to 120g/L
Odor Odorless
Quantity 250 g
UN Number UN1325
Percent Purity ≥99.9%
Chemical Name or Material Carbon black, acetylene, 50% compressed
Hazard Category H351
Hazard Statement GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 215-609-9
RTECSNumber FF5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only