missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbinoxamine Maleate 98.0+%, TCI America™
$124.89 - $408.22
Chemical Identifiers
| CAS | 3505-38-2 |
|---|---|
| Molecular Formula | C20H23ClN2O5 |
| Molecular Weight (g/mol) | 406.863 |
| MDL Number | MFCD00082461 |
| InChI Key | GVNWHCVWDRNXAZ-BTJKTKAUSA-N |
| PubChem CID | 5282409 |
| ChEBI | CHEBI:31353 |
| IUPAC Name | (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-dimethylethanamine |
| SMILES | CN(C)CCOC(C1=CC=C(C=C1)Cl)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O |
Chemical Identifiers
| 3505-38-2 | |
| 406.863 | |
| GVNWHCVWDRNXAZ-BTJKTKAUSA-N | |
| CHEBI:31353 | |
| CN(C)CCOC(C1=CC=C(C=C1)Cl)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O |
| C20H23ClN2O5 | |
| MFCD00082461 | |
| 5282409 | |
| (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-dimethylethanamine |
Specifications
| 3505-38-2 | |
| White | |
| MFCD00082461 | |
| 2811 | |
| CN(C)CCOC(C1=CC=C(C=C1)Cl)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O | |
| 406.863 | |
| CHEBI:31353 | |
| ≥98.0% (HPLC,T) | |
| Carbinoxamine Maleate |
| 119°C | |
| C20H23ClN2O5 | |
| 5 g | |
| GVNWHCVWDRNXAZ-BTJKTKAUSA-N | |
| (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-dimethylethanamine | |
| 5282409 | |
| 406.86 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : US6350000
TSCA : Yes