missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbinoxamine Maleate 98.0+%, TCI America™
Supplier: TCI America C30575G
Specifications
| Carbinoxamine Maleate | |
| 119°C | |
| C20H23ClN2O5 | |
| 5 g | |
| GVNWHCVWDRNXAZ-BTJKTKAUSA-N | |
| (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-dimethylethanamine | |
| 5282409 | |
| 406.86 | |
| Crystalline Powder |
| 3505-38-2 | |
| White | |
| MFCD00082461 | |
| 2811 | |
| CN(C)CCOC(C1=CC=C(C=C1)Cl)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O | |
| 406.863 | |
| CHEBI:31353 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 3505-38-2 | |
| 406.863 | |
| GVNWHCVWDRNXAZ-BTJKTKAUSA-N | |
| CHEBI:31353 | |
| CN(C)CCOC(C1=CC=C(C=C1)Cl)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O |
| C20H23ClN2O5 | |
| MFCD00082461 | |
| 5282409 | |
| (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-dimethylethanamine |
Safety and Handling
RTECSNumber : US6350000
TSCA : Yes