accessibility menu, dialog, popup

UserName

CAS RN 3505-38-2

H3CNCH3OClNOHOOHO(Z)

CAS RN 3505-38-2

IUPAC Name: (2Z)-but-2-enedioic acid; {2-[(4-chlorophenyl)(pyridin-2-yl)methoxy]ethyl}dimethylamine
Synonyms: p-carbinoxamine maleate paracarbinoxamine maleate
Molecular Weight (g/mol): 406.86
Molecular Formula: C20H23ClN2O5
InChi Key: GVNWHCVWDRNXAZ-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CN(C)CCOC(C1=CC=C(Cl)C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results