Learn More
Thermo Scientific™ CaptureSelect™ LambdaXP Affinity Matrix
Thermo Scientific™ CaptureSelect™ LambdaXP Affinity Matrix was specifically designed for the purification of recombinant human immunoglobulins and Fab fragments containing a lambda light chain.
$75427.58 - $75427.58
Specifications
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
|---|---|
| Matrix | Agarose-based, epoxide-activated |
| Form | Agarose Bead |
| For Use With (Application) | Lambda Fab purification |
| Target | Lambda light chain |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
19-437-5201L
|
Thermo Scientific™
194375201L |
1 L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CaptureSelect LambdaXP Affinity Matrix was specifically designed for the purification of recombinant human immunoglobulins and Fab fragments containing a lambda light chain. The ligand binds to the constant domain of the human lambda light chain and recognizes all isotypes including IgA, IgM, IgD, and IgE.
Features of this affinity matrix include:
- Efficient elution of Fab fragments and immunoglobulins containing a lambda light chain, no cross binding to the Fc-region or kappa light chains
- One-step purification from crude material with high purity and yield
- Excellent scalability
- Non-animal-derived
CaptureSelect LambdaXP Affinity Matrix can be utilized for the purification of human Ig's and Fab/Fab2 fragments directly from complex source materials and enables a robust and efficient purification process with excellent purity obtained in a single step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics:
- Matrix: agarose-based, epoxide-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect LambdaXP
- Ligand coupling method: epoxide coupling
- Binding capacity: >30 g IgG/L resin depending on flow rate, column height, and contact time
- Elution conditions: 25 mM sodium acetate, pH 3.5
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
| 50 to 200 cm/hr. (Up to 2 bar) | |
| Agarose Bead | |
| Lambda light chain |
| Agarose-based, epoxide-activated | |
| Lambda Fab purification | |
| LambdaXP |