Learn More
Thermo Scientific™ CaptureSelect™ IgM Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ IgM Ligand Leakage ELISA is designed to detect possible leached ligand of IgM purified using Thermo Scientific™ CaptureSelect™ IgM Affinity Matrix.
$3549.25 - $3549.25
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
|---|---|
| Assay | ELISA |
| For Use With (Equipment) | Microplate Reader |
| Product Line | CaptureSelect |
| Type | ELISA Kit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
81-028-9001
|
Thermo Scientific™
810289001 |
1 Assay |
Each for $3,549.25
|
|
|||||
Description
CaptureSelect IgM Ligand Leakage ELISA is designed to detect possible leached ligand of IgM purified using CaptureSelect IgM Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IgM Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
| Store in freezer (-5°C to -30°C). | |
| Microplate Reader | |
| ELISA Kit |
| ELISA | |
| CaptureSelect | |
| Kit |