missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgM Ligand Leakage ELISA
GREENER_CHOICE

Catalog No. 810289001
Change view
Click to view available options
Quantity:
1 Assay
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
810289001 1 Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 810289001 Supplier Thermo Scientific™ Supplier No. 810289001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ IgM Ligand Leakage ELISA is designed to detect possible leached ligand of IgM purified using Thermo Scientific™ CaptureSelect™ IgM Affinity Matrix.

CaptureSelect IgM Ligand Leakage ELISA is designed to detect possible leached ligand of IgM purified using CaptureSelect IgM Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IgM Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Assay ELISA
For Use With (Equipment) Microplate Reader
Product Line CaptureSelect
Quantity 1 Assay
Type ELISA Kit
Format Kit