Learn More
Thermo Scientific™ CaptureSelect™ IgE Affinity Matrix
Thermo Scientific™ CaptureSelect™ IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.
$75427.58 - $75427.58
Specifications
| Binding Capacity | >5 mg/mL resin |
|---|---|
| Flow Rate | 50 to 200 cm/hr. (Up to 2 bar) |
| Matrix | Agarose-based, epoxide-activated |
| Particle Size | 65 ±10 μm |
| Product Line | CaptureSelect |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
19-435-4201L
|
Thermo Scientific™
194354201L |
1 L |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
CaptureSelect IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.
Features of this affinity matrix include:
- Purification of human IgE, binding different recombinant IgE forms
- Efficient removal of host cell proteins (HCP) and IgE aggregates
- Excellent scalability
- Non–animal-derived
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics
- Matrix: agarose-based, epoxide-activated
- Average particle size: 65 ± 10 μm
- Ligand: CaptureSelect IgE affinity ligand
- Ligand coupling method: aldehyde coupling
- Binding capacity: >5 mg/mL resin
- Elution conditions: 50 mM sodium citrate, 150 mM NaCl, pH 3.5
- Flow characteristics: 50–200 cm/h (up to 2 bar)
- Formulation buffer: 20% (v/v) ethanol
Specifications
| >5 mg/mL resin | |
| Agarose-based, epoxide-activated | |
| CaptureSelect | |
| IgE affinity ligand |
| 50 to 200 cm/hr. (Up to 2 bar) | |
| 65 ±10 μm | |
| Affinity Matrix | |
| IgE |