missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgE Affinity Matrix
GREENER_CHOICE

Catalog No. 194354201l
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Catalog No. Quantity
194354201L 1 L
1 options

Catalog No. 194354201L

Supplier: Thermo Scientific™ 194354201L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.

CaptureSelect IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.

Features of this affinity matrix include:

  • Purification of human IgE, binding different recombinant IgE forms
  • Efficient removal of host cell proteins (HCP) and IgE aggregates
  • Excellent scalability
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect IgE affinity ligand
  • Ligand coupling method: aldehyde coupling
  • Binding capacity: >5 mg/mL resin
  • Elution conditions: 50 mM sodium citrate, 150 mM NaCl, pH 3.5
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity >5 mg/mL resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, epoxide-activated
Particle Size 65 ±10 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 1 L
Type Affinity Matrix
Ligand Type IgE affinity ligand
Stationary Phase IgE