Learn More
Thermo Scientific™ CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix
Thermo Scientific™ CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA.
Supplier:  Thermo Scientific™ 194311010
 View more versions of this product
Catalog No. 19-431-1010
Only null left
Explore available promotions
Description
Description
CaptureSelect IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA. It shows no binding to other species IgA or other immunoglobulins.
This product can be used for purification of:
  • IgA1 and IgA2 from plasma and serum
  • Mono- and dimeric IgA, including secretory IgA
  • IgA Fab fragments
Specifications
Specifications
Store at 2°C to 8°C.
CaptureSelect
Affinity Matrix
Liquid Suspension
10 mL
Anti-IgA-CH1 (Hu)
Product Suggestions
Product Suggestions