missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CaptureSelect™ IgA-CH1 (Hu) Affinity Matrix

Description
CaptureSelect IgA-CH1 (Hu) Affinity Matrix can be used to purify human IgA. It shows no binding to other species IgA or other immunoglobulins.
This product can be used for purification of:
- IgA1 and IgA2 from plasma and serum
- Mono- and dimeric IgA, including secretory IgA
- IgA Fab fragments
Specifications
Specifications
| Content And Storage | Store at 2°C to 8°C. |
| Form | Liquid Suspension |
| Product Line | CaptureSelect |
| Quantity | 5 mL |
| Type | Affinity Matrix |
| Stationary Phase | Anti-IgA-CH1 (Hu) |