missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ HRP Anti-AAVX Conjugate
GREENER_CHOICE

Catalog No. 7303522500
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7303522500 500 μL
7303522100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7303522500 Supplier Thermo Scientific™ Supplier No. 7303522500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ HRP Anti-AAVX Conjugate includes a 14-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of multiple AAV serotypes.

CaptureSelect HRP Anti-AAVX Conjugate includes a 14-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of multiple AAV serotypes. The antibody fragment binds with high affinity to AAV particles and demonstrates good binding reactivity towards a variety of AAV serotypes, including AAV1 to AAV8, and AAVrh10. The antibody fragment is chemically conjugated to horseradish peroxidase (HRP) via a spacer of appropriate length to maintain full binding capacity of the antibody fragment.

CaptureSelect HRP Anti-AAVX Conjugate can be combined with CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522100) in enzyme linked immunoassays to quantify the total number of AAV capsids. Sandwich ELISAs can be designed using the biotin anti-AAVX conjugate as a capture reagent immobilized on a streptavidin coated surface. HRP anti-AAVX conjugate is then used for detection of the bound AAV capsids. The unique binding properties of this pair of anti-AAVX conjugates can circumvent the need for incorporation of different AAV serotype-specific antibodies for AAV assay development.

For assay development to measure AAV9 total capsids, we recommend CaptureSelect Biotin Anti-AAV9 Conjugate (Cat. No. 7103332100) and CaptureSelect HRP Anti-AAV9 Conjugate (Cat. No. 7303332100).

Product details

  • Format type: HRP, liquid
  • Buffer: PBS, 50% (v/v) glycerol
  • Storage: in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.1 mL
  • Recommended working dilution for sandwich ELISA: 1: 5000 to 1: 10,000 (the optimal working dilution should be determined empirically for each application)

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Concentration 0.5 mg/mL
Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 500 μL
For Use With (Application) Quantitation of multiple AAV capsids
Type Conjugate