missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ FcXL Affinity Matrix DFS Item

Thermo Scientific™ CaptureSelect™ FcXL Affinity Matrix has been specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma derived IgG.

Supplier:  Thermo Scientific™ 1943280250

 View more versions of this product

Catalog No. 361054481

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

CaptureSelect FcXL Affinity Matrix has been specifically designed for the purification of recombinant human IgG, Fc fusion proteins, and plasma derived IgG. It recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4).

Features of this affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • Human specific, no binding to bovine antibodies, for example
  • Excellent scalability
  • Non-animal-derived

CaptureSelect FcXL Affinity Matrix purifies IgGs and Fc fusion proteins directly from complex source materials in a single step with high purity and yield.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main Characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect FcXL affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: 25–35 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4, or 20 mM sodium acetate, 1.0 M MgCl2, 40% (v/v) propylene glycol, pH 5–6
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
Specifications

Specifications

25 to 35 g IgG/L
Agarose-based, aldehyde-activated
65 ±10 μm
CaptureSelect
IgG purification
Affinity Matrix
Anti-IgG-Fc
50 to 200 cm/hr. (Up to 2 bar)
Store in refrigerator (2°C to 8°C).
Agarose Bead
250 mL
Fc of IgG, CH3 domain
FcXL affinity ligand
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions