missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-LC-lambda (Human) Conjugate
GREENER_CHOICE

Catalog No. 7103082500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7103082500 500 μg
7103082100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7103082500 Supplier Thermo Scientific™ Supplier No. 7103082500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human lambda light chains.

CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the constant domain of human lambda light chains. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate allows you to:

  • Detect, quantitate, and characterize all human antibody isotypes and subclasses (for example, IgG1 to 4, IgM, IgA, IgD, and IgE), and Fab fragments thereof, that contain a lambda light chain; as well as recombinant human Fab fragments and/or Fab and Fab2 fragments obtained after papain or pepsin digestion
  • Avoid cross-binding with human antibodies that possess a kappa light chain or IgG from, for example, mouse, rat, or rabbit
  • Screen antibody-antigen interactions—biotin anti-LC-lambda (Hu) forms a stable [low k(diss)] complex with, for example, human IgG antibodies through bivalent binding

Applications for CaptureSelect Biotin Anti-LC-lambda (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Form Liquid
Product Line CaptureSelect
Quantity 500 μg
Target Lambda Light Chain
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody