missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG4 (Human) Conjugate
GREENER_CHOICE

Catalog No. 7102902500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7102902500 500 μg
7102902100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7102902500 Supplier Thermo Scientific™ Supplier No. 7102902500
Get an Extra Analytical Consumable with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG4 (Hu) Conjugate consists of a 12 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG4 subclass.

CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate consists of a 12 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG4 subclass. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate allows you to:

  • Detect, quantitate, and characterize human IgG4 antibodies
  • Avoid cross-binding with other IgG subclasses like IgG1, 2 and 3 and isotypes like IgM and IgA including IgG from bovine sources such as FCS

Applications for CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Product Type Biotin Anti-IgG4 Conjugate
Conjugate Biotin and Other Haptens
Quantity 500 μg
Product Line CaptureSelect
Content And Storage Store in freezer (-5°C to -30°C).