missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-Fibrinogen Conjugate
GREENER_CHOICE

Catalog No. 7102912500
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7102912500 500 μg
7102912100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7102912500 Supplier Thermo Scientific™ Supplier No. 7102912500

Thermo Scientific™ CaptureSelect™ Biotin Anti-Fibrinogen Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human fibrinogen.

CaptureSelect Biotin Anti-Fibrinogen Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human fibrinogen. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human fibrinogen.

Applications for CaptureSelect Biotin Anti-Fibrinogen Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, MSIA Streptavidin D.A.R.T.'S, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 500 μg
Type Conjugate
Label or Dye Biotin & Analogs
Ligand Type Antibody