missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ CApture™ BT Fuming System Accessory, Long Gun Holder

Catalog No. 3184100
Change view
Click to view available options
For Use With (Equipment):
CApture BT Fuming System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
3184100 CApture BT Fuming System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3184100 Supplier Labconco™ Supplier No. 3184100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with CApture BT Fuming System

Stainless steel holder provides 5 U-shaped cradles for long guns to be held securely. Holder locks into existing sidewall brackets. CApture BT can accommodate 15 long guns (3 Kits). • Holds 5 long guns securely in U-shaped cradles• Locks into CApture BT's existing sidewall brackets• Stainless steel

Specifications

Product Type Fuming System Chamber
Material Stainless Steel
Dimensions (L x W x H) 68.5 x 15.2 x 3.17 cm (27 x 6 x 1.25 in.)
For Use With (Equipment) CApture BT Fuming System
Includes 5 U-shaped cradles to support long guns
Weight (English) 5 lb.
Weight (Metric) 2 kg