missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals CAPSO sodium salt, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Quantity | 25 g |
| Sensitivity | Hygroscopic |
| Formula Weight | 259.3 |
| Percent Purity | 98% |
| Chemical Name or Material | CAPSO sodium salt |
Sécurité et manipulation
| Hazard Category | H318 |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only