missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Caps for Sample Cups

Designed specifically for use on Globe Scientific multi-purpose sample cups. Globe Scientific Caps for Sample Cups are made from polyethylene and snap securely into place. Caps are available with a solid or cross-cut pierceable top.

Specifications

Color Natural
Material Polyethylene
Quantity 1000/Pk.
For Use With (Equipment) Globe Scientific Sample Cups
Product Type Snap Cap
View More Specs

Products 2
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
361015320
View Documents Promotion Details
Globe Scientific
113132
Flat Top N/A
Only null left
 
21200101
View Documents Promotion Details
Globe Scientific
GLB113133
Pierceable Cross-cut N/A
Only null left
 
Description

Description

  • For use on Globe Scientific sample cups
  • Snap easily and securely into place
  • Available with flat top (21-200-100) or pierceable cross cut (21-200-101)
  • Polyethylene (PE)
  • Natural color
Specifications

Specifications

Natural
1000/Pk.
Snap Cap
Polyethylene
Globe Scientific Sample Cups
Videos
SDS
Documents

Documents