missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Caps for Sample Cups

Catalog No. 21200101
Change view
Click to view available options
Closure Type:
Flat Top
Pierceable Cross-cut
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
21200101 Pierceable Cross-cut
361015320 Flat Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21200101 Supplier Globe Scientific Supplier No. GLB113133

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed specifically for use on Globe Scientific multi-purpose sample cups. Globe Scientific Caps for Sample Cups are made from polyethylene and snap securely into place. Caps are available with a solid or cross-cut pierceable top.

  • For use on Globe Scientific sample cups
  • Snap easily and securely into place
  • Available with flat top (21-200-100) or pierceable cross cut (21-200-101)
  • Polyethylene (PE)
  • Natural color

Specifications

Closure Type Pierceable Cross-cut
Quantity 1000/Pk.
Color Natural
Product Type Snap Cap
Material Polyethylene
For Use With (Equipment) Globe Scientific Sample Cups