missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Caps for Nunclon™ Delta Flasks

Catalog No. 12567251
Change view
Click to view available options
For Use With (Equipment):
175 cm² EasY Flask
80 cm² Nunclon Delta Flask
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
12567251 80 cm² Nunclon Delta Flask
12565626 175 cm² EasY Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12567251 Supplier Thermo Scientific™ Supplier No. 144458

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.

Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close (solid) and filter—to meet your application needs.

Vent/close Caps
  • EasY Flask vent/close caps feature a "Y" mark that allows visual verification
  • Vent position easily distinguised even what flasks are stacked in incubators
Filter Caps
  • Filter membrane provides effective barrier against microorganisms
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
Certifications:
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
Ordering Information:
  • Case of 100

Specifications

Closure Color Blue
Description Sterile, HDPE closure
Material HDPE
For Use With (Equipment) 80 cm² Nunclon Delta Flask
No. per Case 100
No. per Pack 1
Product Type Vent/close cap
Sterility Sterile
Class Class VI
Closure Type Solid
Culture Area 80 cm2
Surface Treatment Nunclon Delta
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.