Learn More
Microsolv Technology Caps Autosampler 9Mm Sil/Ptfe DFS Item
Supplier:  Microsolv Technology Corporation 9502S10CBM
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS.
Catalog No. 502765
Only null left
Explore available promotions