Learn More
Microsolv Technology Corporation Caps Autosam 9Mm Sil/Ptfe Psc
Supplier:  Microsolv Technology Corporation 9502S30CBM
Encompass
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS
Catalog No. 502771
Only null left