missing translation for 'onlineSavingsMsg'
Learn More

ε-Caprolactam, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012383
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012383 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012383 Supplier Merck Emd Millipore Supplier No. 77403100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ε-Caprolactam is used in the manufacture of polyamides. It is also used to produce nylon 6.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Ignition Point 707°F
CAS 105-60-2
Vapor Pressure <0.01 mmHg (20°C)
Flash Point 152°C - Closed Cup
Physical Form Neat
Linear Formula C6H11 NO
MDL Number MFCD00006936
Melting Point 68°C to 71°C
Boiling Point 136°C to 138°C (10 mmHg)
Synonym 2-Oxohexamethyleneimine; Aza-2-cycloheptanone
Type Analytical Standard
Quantity 100 mg
Formula Weight 113.16
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less