missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580946
Description
This antibody has specificity for Human CAP1.
Adenylyl cyclase-associated protein 1 is an enzyme that in humans is encoded by the CAP1 gene. This gene is mapped to 1p34. 2. The protein encoded by this gene is related to the S. cerevisiae CAP protein, which is involved in the cyclic AMP pathway. And the human protein is able to interact with other molecules of the same protein, as well as with CAP2 and actin. Alternatively spliced transcript variants have been identified.Identifiants chimiques
| CAP1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| CAP1 | |
| adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p | |
| CAP1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 1 mg/mL | |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q01518 | |
| 10487 | |
| A synthetic peptide corresponding to the center region of the Human CAP1. | |
| Antibody |
Spécifications
| CAP1 | |
| Polyclonal | |
| Unconjugated | |
| CAP1 | |
| adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10487 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q01518 | |
| CAP1 | |
| A synthetic peptide corresponding to the center region of the Human CAP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |