missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Camphorquinone, 99%

Catalog No. AAA1496706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1496706 5 g
AAA1496714 25 g
AAA1496722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1496706 Supplier Thermo Scientific Chemicals Supplier No. A1496706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10373-78-1 | C10H14O2 | 166.22 g/mol

Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

Solubility
Soluble in ethanol, ethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10373-78-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160
InChI Key VNQXSTWCDUXYEZ-UHFFFAOYSA-N
Synonym camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
PubChem CID 25208
ChEBI CHEBI:34607
IUPAC Name 4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C2CCC1(C(=O)C2=O)C)C

Specifications

Melting Point 198°C to 202°C
Quantity 5 g
Beilstein 1909463
Solubility Information Soluble in ethanol,ethyl ether and benzene.
Formula Weight 166.22
Percent Purity 99%
Chemical Name or Material (+/-)-Camphorquinone
EINECSNumber 233-814-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only