missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ cAMP Recombinant Rabbit Monoclonal Antibody (1Y8W4)
GREENER_CHOICE

Numéro de catalogue. MA569642
Modifier l'affichage
Click to view available options
Quantité:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MA569642 100 μL
MA569643 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA569642 Fournisseur Invitrogen™ Code fournisseur MA569642

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Recombinant Monoclonal Antibody

cAMP Recombinant Monoclonal Antibody for ELISA, IHC (P), Western Blot

Cyclic adenosine monophosphate (cAMP) plays a key role as an intracellular second messenger for transduction events that follow a number of extracellular signals. The G-Protein Coupled Receptors (GPCR) is the largest family of cell surface receptors. They can be activated by different ligands, such as neurotransmitters, hormones, ions, small molecules, peptides, and other physiological signaling molecules. Typically, the binding of the ligands to its receptor resulting in the activation of G-proteins, in return, activates the effector adenylyl cyclase evoking the production of cAMP. The activation of a protein kinase by cAMP results in the phosphorylation of substrate proteins. Currently successful drugs in marketing have been developed to target these receptors. Among the GPCRs, ~367 receptors are potential drug development targets, but only about 20 have been used to generate therapeutically and commercially successful drugs so far. Because the involvement of cAMP can amplify the response of the ligand binding, the second messenger cAMP has been largely employed to monitor the activation of the GPCR to facilitate the therapeutic drug discovery.

Spécifications

Antigène cAMP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1Y8W4
Concentration 1.25 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.09% sodium azide; pH 7.3.
Gène CAMP
Numéro d’acquisition du gène P49913
Alias du gène 18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAMP; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; Cramp; Cyclic AMP; FALL39; FALL-39; FALL-39 peptide antibiotic; HSD26; LL37; MCLP
Symboles de gène CAMP
Espèces hôtes Rabbit
Immunogène Synthetic peptide
Méthode de purification Affinity chromatography
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 820
Espèces cibles Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats