missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Camostat mesylate

Catalog No. 319310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
319310 10 mg
319350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 319310 Supplier Tocris Bioscience™ Supplier No. 3193/10

Orally active protease inhibitor

Camostat mesylate is an orally active protease inhibitor. Inhibits trypsin and various inflammatory proteases including plasmin, kallikrein and thrombin. Suppresses pancreatitis-induced pain in rats following oral administration. Also inhibits transmembrane serine protease TMPRSS2 and partially blocks entry of SARS-Cov-2 (COVID-19) into lung cells in vitro.

Tocris products are for biomedical research use only. They are not intended for human or veterinary use.

Chemical Identifiers

CAS 59721-29-8
Molecular Formula C21H26N4O8S

Specifications

Quantity 10 mg
Formula Weight 494.52
Purity >98%
Molecular Weight (g/mol) 494.52
Chemical Name or Material Camostat mesylate
Recommended Storage Desiccate at Room Temperature