accessibility menu, dialog, popup

UserName

CAS RN 59721-29-8

IUPAC Name: 4-{2-[(dimethylcarbamoyl)methoxy]-2-oxoethyl}phenyl 4-[(diaminomethylidene)amino]benzoate; methanesulfonic acid
Synonyms: camostat; methanesulfonic acid
Molecular Weight (g/mol): 494.52
Molecular Formula: C21H26N4O8S
InChi Key: FSEKIHNIDBATFG-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CN(C)C(=O)COC(=O)CC1=CC=C(OC(=O)C2=CC=C(C=C2)N=C(N)N)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)