missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CaMKI Recombinant Rabbit Monoclonal Antibody (23GB1290)
GREENER_CHOICE

Numéro de catalogue. MA553152
Modifier l'affichage
Click to view available options
Quantité:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MA553152 100 μL
MA553153 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA553152 Fournisseur Invitrogen™ Code fournisseur MA553152
Il en reste null

Rabbit Recombinant Monoclonal Antibody

CaMKI Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

CAMK1 (CaMK1) is a serine/threonine kinase that is a member of the calcium/calmodulin-dependent protein kinase family. It is involved in cell differentiation, nervous system development, and signal transduction. CAMK1 is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.

Spécifications

Antigène CaMKI
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1290
Concentration 0.59 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gène CAMK1
Numéro d’acquisition du gène Q14012, Q63450, Q91YS8
Alias du gène AI505105; calcium/calmodulin dependent protein kinase I; calcium/calmodulin-dependent protein kinase I; calcium/calmodulin-dependent protein kinase type 1; CaM kinase I; CaM kinase I alpha; Camk1; CAMK1-PEN; Camki; caM-KI; CaMKIalpha; CaMKI-alpha; D6Ertd263e; G alpha interacting protein; Gaip; MGC120317; MGC120318; regulator of G-protein signalling 19
Symboles de gène CAMK1
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from human CaMKI.
Méthode de purification Affinity chromatography
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 171503, 52163, 8536
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats