Learn More
Laxco™ Camera C-Mount Adapter
Description
- 0.5X C-mount adapter for mounting CCD/CMOS camera's to Laxco branded microscopes
- Works with all LMC series stands, and Z400/Z230P series stereo heads
Spécifications
Spécifications
| Type | Adapter |
| Magnification Power | 0.5X |
| Material | Rubber |
| Warranty | 5 years optical and mechanical, 1 year electrical from date of purchase |