missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmagite Indicator, 0.1% (w/v), Ricca Chemical
$74.00 - $129.77
Chemical Identifiers
| CAS | 3147-14-6 |
|---|---|
| Molecular Formula | C17H14N2O5S |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | ASFVMSDYPYMUFL-UHFFFAOYSA-N |
| PubChem CID | 6364506 |
| IUPAC Name | 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid |
| SMILES | CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O |
Chemical Identifiers
| 3147-14-6 | |
| Mixture | |
| 6364506 | |
| CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O |
| C17H14N2O5S | |
| ASFVMSDYPYMUFL-UHFFFAOYSA-N | |
| 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid |
Specifications
| 3147-14-6 , 7732-18-5 | |
| 100.0,0.1 | |
| C17H14N2O5S | |
| CC1=CC(=C(C=C1)O)NN=C2C3=CC=CC=C3C(=CC2=O)S(=O)(=O)O | |
| Mixture | |
| 0.1% W/V | |
| Natural Poly Bottle | |
| Red | |
| Liquid |
| 98.0,0.1 | |
| 0.1% (w/v) | |
| ASFVMSDYPYMUFL-UHFFFAOYSA-N | |
| 4-[(2-hydroxy-5-methylphenyl)hydrazinylidene]-3-oxonaphthalene-1-sulfonic acid | |
| 6364506 | |
| Indicator | |
| Aqueous Solution | |
| 120 mL | |
| Calmagite Indicator |