missing translation for 'onlineSavingsMsg'
Learn More

Calix[4]arene, 98%

Catalog No. AAB2132006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2132006 5 g
AAB2132003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2132006 Supplier Thermo Scientific Chemicals Supplier No. B2132006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74568-07-3 | C28H24O4 | 424.496 g/mol

Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,

Solubility
Soluble in chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 74568-07-3
Molecular Formula C28H24O4
Molecular Weight (g/mol) 424.496
MDL Number MFCD00143912
InChI Key YPNHVQZZPXPQOS-UHFFFAOYSA-N
Synonym calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc
PubChem CID 562409
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O

Specifications

Melting Point 315°C to 318°C
Quantity 5 g
Solubility Information Soluble in chloroform.
Formula Weight 424.5
Percent Purity 98%
Chemical Name or Material Calix[4]arene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only