missing translation for 'onlineSavingsMsg'
Learn More

Calix[4]arene, 98%

Catalog No. AC290190050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290190050 5 g Glass bottle
AC290190010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC290190050 Supplier Thermo Scientific Chemicals Supplier No. 290190050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74568-07-3 | C28H24O4 | 424.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 74568-07-3
Molecular Formula C28H24O4
Molecular Weight (g/mol) 424.5
MDL Number MFCD00233673
InChI Key YPNHVQZZPXPQOS-UHFFFAOYSA-N
Synonym calix 4 arene, tetrahydroxycalix 4 arene, calix 4 arene 1g, calix 4 arene-25,26,27,28-tetrol, 25,26,27,28-tetrahydroxycalix 4 aren, 25,26,27,28-tetrahydroxycalix 4 arene, calix 4 arene-25,26,27,28-tetrol, purum hplc
PubChem CID 562409
SMILES C1C2=C(C(=CC=C2)CC3=C(C(=CC=C3)CC4=CC=CC(=C4O)CC5=CC=CC1=C5O)O)O

Specifications

Melting Point 315.0°C to 318.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 424.5
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Calix[4]arene, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only